Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTAAATTCTAGGGCCCAACAGA[C/G]AGAGTGTCTTCATTGCCACCCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613842 MIM: 611270 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GZF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GZF1 - GDNF inducible zinc finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317012.1 | 2702 | Intron | NP_001303941.1 | |||
NM_001317019.1 | 2702 | Intron | NP_001303948.1 | |||
NM_022482.4 | 2702 | Intron | NP_071927.1 |
NAPB - NSF attachment protein beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283018.1 | 2702 | UTR 3 | NP_001269947.1 | |||
NM_001283020.1 | 2702 | UTR 3 | NP_001269949.1 | |||
NM_001283026.1 | 2702 | UTR 3 | NP_001269955.1 | |||
NM_022080.2 | 2702 | UTR 3 | NP_071363.1 | |||
XM_011529313.1 | 2702 | UTR 3 | XP_011527615.1 | |||
XM_011529315.2 | 2702 | Intron | XP_011527617.1 | |||
XM_017028008.1 | 2702 | UTR 3 | XP_016883497.1 |