Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCCCCATTGAGCTGTCTCATGC[A/G]CAGGTTAATGGAGCCGTAGGTCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PSMF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSMF1 - proteasome inhibitor subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM74B - transmembrane protein 74B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304748.1 | 1529 | Missense Mutation | CGC,TGC | R,C 226 | NP_001291677.1 | |
NM_001304749.1 | 1529 | Missense Mutation | CGC,TGC | R,C 202 | NP_001291678.1 | |
NM_018354.2 | 1529 | Missense Mutation | CGC,TGC | R,C 226 | NP_060824.1 | |
XM_011529281.2 | 1529 | Missense Mutation | CGC,TGC | R,C 283 | XP_011527583.1 | |
XM_011529282.2 | 1529 | Missense Mutation | CGC,TGC | R,C 226 | XP_011527584.1 | |
XM_011529283.2 | 1529 | Missense Mutation | CGC,TGC | R,C 226 | XP_011527585.1 | |
XM_011529284.2 | 1529 | Missense Mutation | CGC,TGC | R,C 226 | XP_011527586.1 | |
XM_011529285.2 | 1529 | Missense Mutation | CGC,TGC | R,C 202 | XP_011527587.1 | |
XM_017027923.1 | 1529 | Missense Mutation | CGC,TGC | R,C 258 | XP_016883412.1 | |
XM_017027924.1 | 1529 | Missense Mutation | CGC,TGC | R,C 226 | XP_016883413.1 | |
XM_017027925.1 | 1529 | Missense Mutation | CGC,TGC | R,C 202 | XP_016883414.1 |