Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCTCAGATCCTGGGCTTTTCCA[C/G]CCCTCCAGGCCGGCTCTCCATGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612979 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYS1 - Sys1 golgi trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099791.2 | 398 | Missense Mutation | ACC,AGC | T,S 60 | NP_001093261.1 | |
NM_001197129.1 | 398 | Missense Mutation | ACC,AGC | T,S 60 | NP_001184058.1 | |
NM_033542.3 | 398 | Missense Mutation | ACC,AGC | T,S 60 | NP_291020.1 |
SYS1-DBNDD2 - SYS1-DBNDD2 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TP53TG5 - TP53 target 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |