Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGGAGTCTCAGGAGGATTACG[A/T]TCCTTGCTGGTCCTTGCAGGAGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608377 | ||||||||||||||||||||
Literature Links: |
ARFGAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARFGAP1 - ADP ribosylation factor GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281482.1 | 487 | Missense Mutation | GAT,GTT | D,V 94 | NP_001268411.1 | |
NM_001281483.1 | 487 | Missense Mutation | GAT,GTT | D,V 41 | NP_001268412.1 | |
NM_001281484.1 | 487 | Missense Mutation | ATC,TTC | I,F 15 | NP_001268413.1 | |
NM_018209.3 | 487 | Missense Mutation | GAT,GTT | D,V 94 | NP_060679.1 | |
NM_175609.2 | 487 | Missense Mutation | GAT,GTT | D,V 94 | NP_783202.1 | |
XM_006723823.2 | 487 | Missense Mutation | GAT,GTT | D,V 94 | XP_006723886.1 | |
XM_011528901.1 | 487 | Missense Mutation | GAT,GTT | D,V 94 | XP_011527203.1 | |
XM_017027950.1 | 487 | Missense Mutation | GAT,GTT | D,V 94 | XP_016883439.1 |