Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATAATCTCCTGTAACCTTTTGT[C/T]TTCATGTGATTGCAGTCACCAAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607167 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYNLRB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DYNLRB1 - dynein light chain roadblock-type 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281727.1 | Intron | NP_001268656.1 | ||||
NM_001281728.1 | Intron | NP_001268657.1 | ||||
NM_001281729.1 | Intron | NP_001268658.1 | ||||
NM_001319157.1 | Intron | NP_001306086.1 | ||||
NM_014183.3 | Intron | NP_054902.1 | ||||
NM_177953.2 | Intron | NP_808852.1 |