Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGGTGCCCGTCATCATCCACCG[C/T]GACAACATCATCAGTGACTATGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GDAP1L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GDAP1L1 - ganglioside induced differentiation associated protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256737.1 | 236 | Silent Mutation | CGC,CGT | R,R 104 | NP_001243666.1 | |
NM_001256738.1 | 236 | Silent Mutation | CGC,CGT | R,R 15 | NP_001243667.1 | |
NM_001256739.1 | 236 | Silent Mutation | CGC,CGT | R,R 104 | NP_001243668.1 | |
NM_001256740.1 | 236 | Silent Mutation | CGC,CGT | R,R 104 | NP_001243669.1 | |
NM_024034.4 | 236 | Silent Mutation | CGC,CGT | R,R 104 | NP_076939.3 |