Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTTATTGATGTAGAAATTGCCC[A/G]CAGCGATTGCAGAAAGCCTTTAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608683 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CST8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CST8 - cystatin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281730.1 | 660 | Missense Mutation | CAC,CGC | H,R 92 | NP_001268659.1 | |
NM_005492.3 | 660 | Missense Mutation | CAC,CGC | H,R 92 | NP_005483.1 |