Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGCTGAGCTCCATCTTGTAGTT[A/G]AGCTTCTCCTCCGCCTGGCGGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608362 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STMN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STMN3 - stathmin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276310.1 | 673 | Silent Mutation | CTC,CTT | L,L 125 | NP_001263239.1 | |
NM_015894.3 | 673 | Silent Mutation | CTC,CTT | L,L 136 | NP_056978.2 |