Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATGACGAGGTGCCGGTGGAGCC[A/C]GAGTCTGACTCCGGGGACGAGGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616621 MIM: 601716 | ||||||||||||||||||||
Literature Links: |
DDX27 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX27 - DEAD-box helicase 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017895.7 | 188 | Silent Mutation | CCA,CCC | P,P 52 | NP_060365.7 | |
XM_011528888.2 | 188 | Silent Mutation | CCA,CCC | P,P 52 | XP_011527190.1 |
STAU1 - staufen double-stranded RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037328.2 | 188 | Intron | NP_001032405.1 | |||
NM_001319134.1 | 188 | Intron | NP_001306063.1 | |||
NM_001319135.1 | 188 | Intron | NP_001306064.1 | |||
NM_001322927.1 | 188 | Intron | NP_001309856.1 | |||
NM_001322928.1 | 188 | Intron | NP_001309857.1 | |||
NM_001322929.1 | 188 | Intron | NP_001309858.1 | |||
NM_001322930.1 | 188 | Intron | NP_001309859.1 | |||
NM_001322931.1 | 188 | Intron | NP_001309860.1 | |||
NM_001322932.1 | 188 | Intron | NP_001309861.1 | |||
NM_001322933.1 | 188 | Intron | NP_001309862.1 | |||
NM_004602.3 | 188 | Intron | NP_004593.2 | |||
NM_017452.3 | 188 | Intron | NP_059346.2 | |||
NM_017453.3 | 188 | Intron | NP_059347.2 | |||
NM_017454.3 | 188 | Intron | NP_059348.2 | |||
XM_005260527.1 | 188 | Intron | XP_005260584.1 | |||
XM_006723865.1 | 188 | Intron | XP_006723928.1 | |||
XM_011529015.1 | 188 | Intron | XP_011527317.1 | |||
XM_017028028.1 | 188 | Intron | XP_016883517.1 |