Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGTTATATTCTGGCATCTGCTGT[C/T]GGCATCTCTCATTTGATAGAAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607247 MIM: 606635 | ||||||||||||||||||||
Literature Links: |
CHODL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHODL - chondrolectin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMPRSS15 - transmembrane protease, serine 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002772.2 | 3020 | Missense Mutation | CAA,CGA | Q,R 942 | NP_002763.2 | |
XM_011529654.1 | 3020 | Missense Mutation | CAA,CGA | Q,R 987 | XP_011527956.1 | |
XM_011529655.1 | 3020 | Missense Mutation | CAA,CGA | Q,R 987 | XP_011527957.1 | |
XM_011529656.2 | 3020 | Missense Mutation | CAA,CGA | Q,R 987 | XP_011527958.1 | |
XM_011529657.1 | 3020 | Missense Mutation | CAA,CGA | Q,R 972 | XP_011527959.1 | |
XM_011529658.1 | 3020 | Missense Mutation | CAA,CGA | Q,R 960 | XP_011527960.1 | |
XM_011529659.1 | 3020 | Missense Mutation | CAA,CGA | Q,R 957 | XP_011527961.1 |