Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTATCTGCAAGTACAGGTACTAT[A/T]ACTTACACCTTAAAATGCTTTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616020 MIM: 616021 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYYR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYYR1 - cysteine and tyrosine rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320768.1 | 2394 | UTR 3 | NP_001307697.1 | |||
NM_052954.3 | 2394 | UTR 3 | NP_443186.2 | |||
XM_011529450.2 | 2394 | Intron | XP_011527752.1 |
CYYR1-AS1 - cysteine and tyrosine rich 1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |