Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTTTAGTCGTTCGCCTAGACAC[A/G]TTCCTGTCGCCTCTGACAGCTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600909 MIM: 603294 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LSS - lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCM3AP - minichromosome maintenance complex component 3 associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003906.4 | 6411 | Missense Mutation | ACG,ATG | T,M 1944 | NP_003897.2 | |
XM_005261203.4 | 6411 | Missense Mutation | ACG,ATG | T,M 1944 | XP_005261260.1 | |
XM_005261204.4 | 6411 | Missense Mutation | ACG,ATG | T,M 1944 | XP_005261261.1 | |
XM_005261205.3 | 6411 | Missense Mutation | ACG,ATG | T,M 1944 | XP_005261262.1 |
MCM3AP-AS1 - MCM3AP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |