Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATACCTGCGGAGTTCCGGGGAG[C/T]AAGTACTGGCCAGTTTCCCAGTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609892 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RIPPLY3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RIPPLY3 - ripply transcriptional repressor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317768.1 | 502 | Nonsense Mutation | CAA,TAA | Q,* 14 | NP_001304697.1 | |
NM_001317777.1 | 502 | Nonsense Mutation | CAA,TAA | Q,* 14 | NP_001304706.1 | |
NM_018962.2 | 502 | Nonsense Mutation | CAA,TAA | Q,* 98 | NP_061835.1 |