Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTCTGCAGGGCCTTGCGGCCCTC[A/G]GGGGCCTCTGCCAGCATGGTAAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605663 | ||||||||||||||||||||
Literature Links: |
RSPH14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RSPH14 - radial spoke head 14 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014433.2 | 1402 | Silent Mutation | CCC,CCT | P,P 303 | NP_055248.1 | |
XM_011530149.2 | 1402 | Silent Mutation | CCC,CCT | P,P 353 | XP_011528451.1 | |
XM_011530150.1 | 1402 | Nonsense Mutation | CGA,TGA | R,* 348 | XP_011528452.1 | |
XM_011530151.2 | 1402 | Nonsense Mutation | CGA,TGA | R,* 348 | XP_011528453.1 | |
XM_011530152.2 | 1402 | Nonsense Mutation | CGA,TGA | R,* 348 | XP_011528454.1 | |
XM_011530154.2 | 1402 | Nonsense Mutation | CGA,TGA | R,* 280 | XP_011528456.1 | |
XM_011530155.2 | 1402 | Nonsense Mutation | CGA,TGA | R,* 216 | XP_011528457.1 | |
XM_017028774.1 | 1402 | Nonsense Mutation | CGA,TGA | R,* 348 | XP_016884263.1 |