Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTGGGAGAGGATGGACGAGGGA[A/T]GCGGAGAGACCATATATGTCATTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602245 MIM: 615323 | ||||||||||||||||||||
Literature Links: |
GTPBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GTPBP1 - GTP binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004286.4 | 1646 | Missense Mutation | AGC,TGC | S,C 90 | NP_004277.2 | |
XM_011530537.2 | 1646 | Missense Mutation | AGC,TGC | S,C 90 | XP_011528839.1 | |
XM_017029099.1 | 1646 | Missense Mutation | AGC,TGC | S,C 90 | XP_016884588.1 | |
XM_017029100.1 | 1646 | Missense Mutation | AGC,TGC | S,C 90 | XP_016884589.1 | |
XM_017029101.1 | 1646 | Missense Mutation | GAA,GAT | E,D 15 | XP_016884590.1 | |
XM_017029102.1 | 1646 | Intron | XP_016884591.1 | |||
XM_017029103.1 | 1646 | Intron | XP_016884592.1 |
JOSD1 - Josephin domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |