Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACGTGGCCAGCACAGATCTGCAG[C/G]ACGCGCACCAGGCGCTGCGCCATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610152 | ||||||||||||||||||||
Literature Links: |
CENPM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CENPM - centromere protein M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002876.2 | 556 | UTR 3 | NP_001002876.1 | |||
NM_001110215.2 | 556 | Silent Mutation | GTC,GTG | V,V 27 | NP_001103685.1 | |
NM_001304370.1 | 556 | Silent Mutation | GTC,GTG | V,V 115 | NP_001291299.1 | |
NM_001304371.1 | 556 | Intron | NP_001291300.1 | |||
NM_001304372.1 | 556 | UTR 3 | NP_001291301.1 | |||
NM_001304373.1 | 556 | UTR 3 | NP_001291302.1 | |||
NM_024053.4 | 556 | Silent Mutation | GTC,GTG | V,V 149 | NP_076958.1 | |
XM_011530368.2 | 556 | Intron | XP_011528670.1 |