Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCACCGCACGCTAAAGGAGATTC[C/T]CAGGTGAGGGTCTCCCTCTGGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607110 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOBEC3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOBEC3B - apolipoprotein B mRNA editing enzyme catalytic subunit 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270411.1 | 621 | Missense Mutation | CCC,CTC | P,L 189 | NP_001257340.1 | |
NM_004900.4 | 621 | Missense Mutation | CCC,CTC | P,L 189 | NP_004891.4 |
APOBEC3B-AS1 - APOBEC3B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |