Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAAAAAAGCTGGTGGCTGGCTC[A/G]TGGAGGGCCTTGTATATTATGTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611336 MIM: 606796 MIM: 613553 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJB7 - DnaJ heat shock protein family (Hsp40) member B7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145174.1 | Intron | NP_660157.1 |
ST13 - suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XPNPEP3 - X-prolyl aminopeptidase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204827.1 | Intron | NP_001191756.1 | ||||
NM_022098.3 | Intron | NP_071381.1 |