Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGTGGGCCTAAGGGCTGTGCCCCT[A/G]GACCTGGCACAACTGGTTACTACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613441 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC35E4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC35E4 - solute carrier family 35 member E4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001479.3 | 2127 | Silent Mutation | CTA,CTG | L,L 133 | NP_001001479.1 | |
NM_001318370.1 | 2127 | Silent Mutation | CTA,CTG | L,L 133 | NP_001305299.1 | |
NM_001318371.1 | 2127 | Silent Mutation | CTA,CTG | L,L 133 | NP_001305300.1 | |
XM_017028794.1 | 2127 | Silent Mutation | CTA,CTG | L,L 133 | XP_016884283.1 | |
XM_017028795.1 | 2127 | Silent Mutation | CTA,CTG | L,L 133 | XP_016884284.1 |
TCN2 - transcobalamin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |