Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCATCACACTCTGGCTCTGCA[C/T]GATGCTGCGCTCGCTGGGCTGCGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614910 | ||||||||||||||||||||
Literature Links: |
C1QTNF6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1QTNF6 - C1q and tumor necrosis factor related protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031910.3 | 3256 | Missense Mutation | ATG,GTG | M,V 229 | NP_114116.3 | |
NM_182486.1 | 3256 | Missense Mutation | ATG,GTG | M,V 229 | NP_872292.1 | |
XM_011529857.1 | 3256 | Missense Mutation | ATG,GTG | M,V 210 | XP_011528159.1 | |
XM_017028569.1 | 3256 | Missense Mutation | ATG,GTG | M,V 229 | XP_016884058.1 | |
XM_017028570.1 | 3256 | Missense Mutation | ATG,GTG | M,V 210 | XP_016884059.1 | |
XM_017028571.1 | 3256 | Missense Mutation | ATG,GTG | M,V 210 | XP_016884060.1 | |
XM_017028572.1 | 3256 | Missense Mutation | ATG,GTG | M,V 210 | XP_016884061.1 | |
XM_017028573.1 | 3256 | Missense Mutation | ATG,GTG | M,V 210 | XP_016884062.1 | |
XM_017028574.1 | 3256 | Missense Mutation | ATG,GTG | M,V 210 | XP_016884063.1 |