Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCTCAGAGGCAAAGTCCGGGTC[A/G]GCCTGGCTGCGGTCAGGGGTCCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613619 | ||||||||||||||||||||
Literature Links: |
KLHL22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLHL22 - kelch like family member 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032775.3 | 4550 | Silent Mutation | NP_116164.2 | |||
XM_017029018.1 | 4550 | Silent Mutation | XP_016884507.1 | |||
XM_017029019.1 | 4550 | Silent Mutation | XP_016884508.1 | |||
XM_017029020.1 | 4550 | Silent Mutation | XP_016884509.1 | |||
XM_017029021.1 | 4550 | Silent Mutation | XP_016884510.1 | |||
XM_017029022.1 | 4550 | Silent Mutation | XP_016884511.1 | |||
XM_017029023.1 | 4550 | Silent Mutation | XP_016884512.1 | |||
XM_017029024.1 | 4550 | Silent Mutation | XP_016884513.1 | |||
XM_017029025.1 | 4550 | Silent Mutation | XP_016884514.1 |
SCARF2 - scavenger receptor class F member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |