Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACCTAGCCCCAGTTGCACTCAC[A/G]TCCGAGAACCGGGCTGGCTGGGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 170998 | ||||||||||||||||||||
Literature Links: |
CDPF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDPF1 - cysteine rich DPF motif domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207327.4 | 796 | Missense Mutation | ACG,ATG | T,M 123 | NP_997210.3 | |
XM_011529960.1 | 796 | Silent Mutation | GAC,GAT | D,D 124 | XP_011528262.1 | |
XM_011529962.2 | 796 | Silent Mutation | GAC,GAT | D,D 124 | XP_011528264.1 | |
XM_011529963.1 | 796 | Intron | XP_011528265.1 | |||
XM_011529964.1 | 796 | Intron | XP_011528266.1 | |||
XM_011529965.2 | 796 | Missense Mutation | ACG,ATG | T,M 123 | XP_011528267.1 |
PPARA - peroxisome proliferator activated receptor alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |