Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACGGTGTCACTCAGGGACTTAAG[C/T]TGAGCCACTTGCTCCATAAGTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601143 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCTN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCTN1 - dynactin subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135040.2 | 3539 | Silent Mutation | CAA,CAG | Q,Q 1166 | NP_001128512.1 | |
NM_001135041.2 | 3539 | Silent Mutation | CAA,CAG | Q,Q 1052 | NP_001128513.1 | |
NM_001190836.1 | 3539 | Silent Mutation | CAA,CAG | Q,Q 1149 | NP_001177765.1 | |
NM_001190837.1 | 3539 | Silent Mutation | CAA,CAG | Q,Q 1184 | NP_001177766.1 | |
NM_004082.4 | 3539 | Silent Mutation | CAA,CAG | Q,Q 1191 | NP_004073.2 | |
NM_023019.3 | 3539 | Silent Mutation | CAA,CAG | Q,Q 1057 | NP_075408.1 |