Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGACTGAGCAGCCAACAAGGAG[A/G]GCCTGCCAGAACTGAGCACAGCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616010 | ||||||||||||||||||||
Literature Links: |
COPS7B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COPS7B - COP9 signalosome subunit 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282949.2 | 380 | Missense Mutation | AGC,GGC | S,G 50 | NP_001269878.1 | |
NM_001282950.2 | 380 | Missense Mutation | AGC,GGC | S,G 84 | NP_001269879.1 | |
NM_001282951.2 | 380 | Intron | NP_001269880.1 | |||
NM_001282952.2 | 380 | UTR 5 | NP_001269881.1 | |||
NM_001308381.1 | 380 | Missense Mutation | AGC,GGC | S,G 84 | NP_001295310.1 | |
NM_022730.3 | 380 | Missense Mutation | AGC,GGC | S,G 84 | NP_073567.1 | |
XM_006712693.1 | 380 | Missense Mutation | AGC,GGC | S,G 84 | XP_006712756.1 | |
XM_011511638.1 | 380 | UTR 5 | XP_011509940.1 | |||
XM_011511639.1 | 380 | UTR 5 | XP_011509941.1 | |||
XM_011511640.1 | 380 | UTR 5 | XP_011509942.1 | |||
XM_017004716.1 | 380 | Missense Mutation | AGC,GGC | S,G 84 | XP_016860205.1 | |
XM_017004717.1 | 380 | UTR 5 | XP_016860206.1 |