Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGATAAGGTTTTTTTTACAGTCA[C/T]CTGTAAAGTAGACAACCCACATACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606704 MIM: 607705 | ||||||||||||||||||||
Literature Links: |
GPR75 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR75 - G protein-coupled receptor 75 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPR75-ASB3 - GPR75-ASB3 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSME4 - proteasome activator subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014614.2 | 5490 | Missense Mutation | ATG,GTG | M,V 1800 | NP_055429.2 | |
XM_006711969.1 | 5490 | Missense Mutation | ATG,GTG | M,V 958 | XP_006712032.1 | |
XM_011532705.2 | 5490 | Missense Mutation | ATG,GTG | M,V 1826 | XP_011531007.1 | |
XM_011532706.2 | 5490 | Missense Mutation | ATG,GTG | M,V 1822 | XP_011531008.1 | |
XM_011532707.2 | 5490 | Missense Mutation | ATG,GTG | M,V 1804 | XP_011531009.1 | |
XM_011532709.2 | 5490 | Intron | XP_011531011.1 |