Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTTTAAGTATCATTTACACAAT[A/G]AACTCTTAAGTCTTTAAAAAAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608960 MIM: 601949 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL5A - ADP ribosylation factor like GTPase 5A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CACNB4 - calcium voltage-gated channel auxiliary subunit beta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011511795.2 | 5034 | UTR 3 | XP_011510097.1 | |||
XM_011511796.2 | 5034 | UTR 3 | XP_011510098.1 | |||
XM_011511797.2 | 5034 | UTR 3 | XP_011510099.1 | |||
XM_011511798.2 | 5034 | Intron | XP_011510100.1 | |||
XM_017004880.1 | 5034 | UTR 3 | XP_016860369.1 | |||
XM_017004881.1 | 5034 | UTR 3 | XP_016860370.1 | |||
XM_017004882.1 | 5034 | UTR 3 | XP_016860371.1 | |||
XM_017004883.1 | 5034 | UTR 3 | XP_016860372.1 | |||
XM_017004884.1 | 5034 | UTR 3 | XP_016860373.1 | |||
XM_017004885.1 | 5034 | UTR 3 | XP_016860374.1 | |||
XM_017004886.1 | 5034 | UTR 3 | XP_016860375.1 | |||
XM_017004887.1 | 5034 | Intron | XP_016860376.1 | |||
XM_017004888.1 | 5034 | Intron | XP_016860377.1 |