Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCGTGCCTGGCCCAGGAGGGCC[C/T]CCAGGTACGTGTTGGCTCTCTGCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 188855 | ||||||||||||||||||||
Literature Links: |
GNLY PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GNLY - granulysin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302758.1 | 294 | Missense Mutation | CCC,CTC | P,L 78 | NP_001289687.1 | |
NM_006433.4 | 294 | Missense Mutation | CCC,CTC | P,L 51 | NP_006424.2 | |
NM_012483.3 | 294 | Missense Mutation | CCC,CTC | P,L 36 | NP_036615.2 | |
XM_005264084.2 | 294 | Missense Mutation | CCC,CTC | P,L 78 | XP_005264141.1 | |
XM_005264085.2 | 294 | Missense Mutation | CCC,CTC | P,L 51 | XP_005264142.1 | |
XM_005264087.2 | 294 | Missense Mutation | CCC,CTC | P,L 36 | XP_005264144.1 |