Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGGAACCAGATCCTGACCTTCC[A/G]GATTCAGGTATGGCTCCTCCATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FER1L5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FER1L5 - fer-1 like family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293083.1 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | NP_001280012.1 | |
XM_011512110.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | XP_011510412.1 | |
XM_011512111.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | XP_011510413.1 | |
XM_011512113.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | XP_011510415.1 | |
XM_011512114.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | XP_011510416.1 | |
XM_011512115.2 | 1251 | Intron | XP_011510417.1 | |||
XM_011512116.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | XP_011510418.1 | |
XM_011512117.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | XP_011510419.1 | |
XM_011512118.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 338 | XP_011510420.1 | |
XM_011512119.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | XP_011510421.1 | |
XM_011512120.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 308 | XP_011510422.1 | |
XM_011512122.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | XP_011510424.1 | |
XM_011512124.2 | 1251 | Intron | XP_011510426.1 | |||
XM_011512125.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | XP_011510427.1 | |
XM_011512126.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | XP_011510428.1 | |
XM_011512127.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | XP_011510429.1 | |
XM_011512128.2 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | XP_011510430.1 | |
XM_017005222.1 | 1251 | Missense Mutation | CAG,CGG | Q,R 378 | XP_016860711.1 | |
XM_017005223.1 | 1251 | Intron | XP_016860712.1 | |||
XM_017005224.1 | 1251 | Intron | XP_016860713.1 |