Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGATGATGAAGATATTGATTATG[C/T]TTGCTCTTGGAATGAACTACTGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602637 | ||||||||||||||||||||
Literature Links: |
SPP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPP2 - secreted phosphoprotein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006944.2 | 168 | Missense Mutation | CTT,TTT | L,F 18 | NP_008875.1 | |
XM_005246102.3 | 168 | Missense Mutation | CTT,TTT | L,F 18 | XP_005246159.1 | |
XM_011511698.2 | 168 | Missense Mutation | CTT,TTT | L,F 18 | XP_011510000.1 | |
XM_011511699.2 | 168 | Missense Mutation | CTT,TTT | L,F 18 | XP_011510001.1 | |
XM_011511700.2 | 168 | Missense Mutation | CTT,TTT | L,F 18 | XP_011510002.1 |