Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCTTCGGGGGTAACCGCTTCAT[A/G]GGGTGGTGCTCTCCACACAGGGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605538 | ||||||||||||||||||||
Literature Links: |
NTSR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NTSR2 - neurotensin receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012344.3 | 1220 | Silent Mutation | NP_036476.1 | |||
XM_005246156.3 | 1220 | UTR 3 | XP_005246213.1 | |||
XM_006711876.3 | 1220 | UTR 3 | XP_006711939.1 | |||
XM_006711877.3 | 1220 | UTR 3 | XP_006711940.1 | |||
XM_017003738.1 | 1220 | UTR 3 | XP_016859227.1 |