Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGCGCAGTACAGTGCAGTGTGG[C/T]AAAATGACATATGCCTGTATTTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613920 MIM: 604623 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COA5 - cytochrome c oxidase assembly factor 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MGAT4A - mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNC50 - unc-50 inner nuclear membrane RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005263913.4 | Intron | XP_005263970.1 | ||||
XM_006712403.3 | Intron | XP_006712466.1 | ||||
XM_006712404.2 | Intron | XP_006712467.1 |