Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAATTTCCGGCACACCAGCTCC[C/T]CTTGGTAAGGCCACCCCACCCCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612283 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4783 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4783 - microRNA 4783 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PROC - protein C, inactivator of coagulation factors Va and VIIIa | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000312.3 | 83 | Silent Mutation | CCC,CCT | P,P 22 | NP_000303.1 | |
XM_005263717.4 | 83 | Silent Mutation | CCC,CCT | P,P 43 | XP_005263774.1 | |
XM_017004505.1 | 83 | Missense Mutation | CCC,CTC | P,L 115 | XP_016859994.1 | |
XM_017004506.1 | 83 | Silent Mutation | CCC,CCT | P,P 22 | XP_016859995.1 |