Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGACAGGATCAGGATTTGACTCAG[G/T]AGGATTAGAATGTGAAGAATCCGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137167 MIM: 601468 | ||||||||||||||||||||
Literature Links: |
GGCX PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GGCX - gamma-glutamyl carboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000821.6 | 2161 | Missense Mutation | ACT,CCT | T,P 747 | NP_000812.2 | |
NM_001142269.3 | 2161 | Missense Mutation | ACT,CCT | T,P 690 | NP_001135741.1 | |
NM_001311312.1 | 2161 | Intron | NP_001298241.1 | |||
XM_005264259.4 | 2161 | Missense Mutation | ACT,CCT | T,P 745 | XP_005264316.1 | |
XM_011532764.2 | 2161 | Missense Mutation | ACT,CCT | T,P 473 | XP_011531066.1 | |
XM_011532765.2 | 2161 | Missense Mutation | ACT,CCT | T,P 473 | XP_011531067.1 | |
XM_017003803.1 | 2161 | Missense Mutation | ACT,CCT | T,P 688 | XP_016859292.1 |
MAT2A - methionine adenosyltransferase 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |