Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAATATGGTGAGGCCGATCAGAA[C/T]GCCTATGCTGCCGTCCAGGTACCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601774 | ||||||||||||||||||||
Literature Links: |
MGAT5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MGAT5 - mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM163 - transmembrane protein 163 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030923.4 | 944 | Missense Mutation | ATT,GTT | I,V 258 | NP_112185.1 | |
XM_011511950.2 | 944 | Missense Mutation | ATT,GTT | I,V 147 | XP_011510252.1 | |
XM_017005060.1 | 944 | Intron | XP_016860549.1 | |||
XM_017005061.1 | 944 | Missense Mutation | ATT,GTT | I,V 147 | XP_016860550.1 | |
XM_017005062.1 | 944 | Intron | XP_016860551.1 |