Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTGGTGGAGACAAGCGTGAGGC[C/T]CTGTAGCGGGGTCAGGGAAGCCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603202 MIM: 611216 | ||||||||||||||||||||
Literature Links: |
LCT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LCT - lactase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002299.3 | 5548 | Missense Mutation | GAG,GGG | E,G 1846 | NP_002290.2 | |
XM_017004088.1 | 5548 | Intron | XP_016859577.1 |
UBXN4 - UBX domain protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |