Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTCTTCATACTCCTTTTTGGAC[A/G]CCCACATCTTCTTAAAAGTGTCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605144 MIM: 123866 | ||||||||||||||||||||
Literature Links: |
ACTR1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTR1B - ARP1 actin-related protein 1 homolog B, centractin beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005735.3 | 1335 | Missense Mutation | GCG,GTG | A,V 358 | NP_005726.1 | |
XM_005263854.4 | 1335 | Missense Mutation | GCG,GTG | A,V 284 | XP_005263911.1 | |
XM_017003116.1 | 1335 | Missense Mutation | GCG,GTG | A,V 314 | XP_016858605.1 |
COX5B - cytochrome c oxidase subunit 5B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |