Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCGATGCTCAACTCCGGGACAAA[A/G]AGTTTTACAGGCCCATCCCTAACCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC101928371 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101928371 - uncharacterized LOC101928371 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX37 - testis expressed 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152670.2 | 271 | Missense Mutation | AAG,GAG | K,E 52 | NP_689883.1 | |
XM_005264188.3 | 271 | Missense Mutation | AAG,GAG | K,E 52 | XP_005264245.3 | |
XM_011532691.1 | 271 | Missense Mutation | AAG,GAG | K,E 52 | XP_011530993.1 | |
XM_011532692.1 | 271 | Missense Mutation | AAG,GAG | K,E 52 | XP_011530994.1 |