Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGTGGCTAGACTCACGCTTTGTT[A/C]TGGGCGGCGTTGAATGGTGTCCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CLEC4F PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLEC4F - C-type lectin domain family 4 member F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258027.1 | 1727 | Missense Mutation | CAG,CAT | Q,H 550 | NP_001244956.1 | |
NM_001321308.1 | 1727 | Missense Mutation | CAG,CAT | Q,H 550 | NP_001308237.1 | |
NM_173535.2 | 1727 | Missense Mutation | CAG,CAT | Q,H 550 | NP_775806.2 | |
XM_011532635.2 | 1727 | Missense Mutation | CAG,CAT | Q,H 660 | XP_011530937.1 | |
XM_011532637.2 | 1727 | Missense Mutation | CAG,CAT | Q,H 660 | XP_011530939.1 | |
XM_011532638.2 | 1727 | Missense Mutation | CAG,CAT | Q,H 660 | XP_011530940.1 | |
XM_011532639.2 | 1727 | Missense Mutation | CAG,CAT | Q,H 660 | XP_011530941.1 | |
XM_011532640.2 | 1727 | Missense Mutation | CAG,CAT | Q,H 660 | XP_011530942.1 | |
XM_011532641.2 | 1727 | UTR 3 | XP_011530943.1 | |||
XM_011532642.2 | 1727 | Missense Mutation | CAG,CAT | Q,H 520 | XP_011530944.1 | |
XM_011532643.1 | 1727 | Missense Mutation | CAG,CAT | Q,H 489 | XP_011530945.1 | |
XM_017003519.1 | 1727 | Missense Mutation | CAG,CAT | Q,H 660 | XP_016859008.1 |