Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTAGGAGGCCAACTTACCCAGGC[C/T]CTCTGTGGCTTCCACACAGATGCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM228B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM228B - family with sequence similarity 228 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145710.1 | 727 | Intron | NP_001139182.1 | |||
NM_001291328.1 | 727 | Intron | NP_001278257.1 |
PFN4 - profilin family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199346.2 | 727 | Missense Mutation | AGC,GGC | S,G 119 | NP_955378.1 | |
XM_005264292.2 | 727 | Missense Mutation | AGC,GGC | S,G 119 | XP_005264349.1 | |
XM_006712007.3 | 727 | Missense Mutation | AGC,GGC | S,G 119 | XP_006712070.1 | |
XM_011532823.2 | 727 | Missense Mutation | AGC,GGC | S,G 119 | XP_011531125.1 |