Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGGACCAGTAGATTGGAAGGAA[C/T]GATGTGTAGCTCTGGAGTCCCAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612723 | ||||||||||||||||||||
Literature Links: |
PLEKHH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLEKHH2 - pleckstrin homology, MyTH4 and FERM domain containing H2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172069.3 | 129 | Nonsense Mutation | CGA,TGA | R,* 16 | NP_742066.2 | |
XM_017003351.1 | 129 | Intron | XP_016858840.1 | |||
XM_017003352.1 | 129 | Nonsense Mutation | CGA,TGA | R,* 16 | XP_016858841.1 | |
XM_017003353.1 | 129 | UTR 5 | XP_016858842.1 |