Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAAAGTATTTATATAACCATTGA[A/G]GACATGTTTTAATTAAGGAAGCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603839 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM126B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM126B - family with sequence similarity 126 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFB3 - NADH:ubiquinone oxidoreductase subunit B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257102.1 | Intron | NP_001244031.1 | ||||
NM_002491.2 | Intron | NP_002482.1 | ||||
XM_011511230.2 | Intron | XP_011509532.1 | ||||
XM_017004186.1 | Intron | XP_016859675.1 |