Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGAAGAATGGACATTCGTCAGAA[C/T]TGATGCCATCAAGGTGGAAATCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612003 MIM: 603208 | ||||||||||||||||||||
Literature Links: |
GIGYF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GIGYF2 - GRB10 interacting GYF protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001103146.1 | 590 | Intron | NP_001096616.1 | |||
NM_001103147.1 | 590 | Intron | NP_001096617.1 | |||
NM_001103148.1 | 590 | Intron | NP_001096618.1 | |||
NM_015575.3 | 590 | Intron | NP_056390.2 |
KCNJ13 - potassium voltage-gated channel subfamily J member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172416.1 | 590 | UTR 3 | NP_001165887.1 | |||
NM_001172417.1 | 590 | Missense Mutation | AAT,AGT | N,S 150 | NP_001165888.1 | |
NM_002242.4 | 590 | Missense Mutation | AAT,AGT | N,S 230 | NP_002233.2 |