Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCGCCCTCCTCCACTTCCACCA[C/T]GCCTGCAAGGGAAGGGCAGTCAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612533 | ||||||||||||||||||||
Literature Links: |
THAP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
THAP4 - THAP domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164356.1 | 1806 | Missense Mutation | NP_001157828.1 | |||
NM_015963.5 | 1806 | Missense Mutation | NP_057047.4 | |||
XM_005247016.4 | 1806 | Missense Mutation | XP_005247073.3 | |||
XM_011511291.2 | 1806 | Missense Mutation | XP_011509593.1 | |||
XM_017004256.1 | 1806 | Missense Mutation | XP_016859745.1 |