Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCCTGTAGACCCAGGCGAAGAT[A/G]TTGGGGCTCACGTTGTGGTCGGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612930 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PID1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PID1 - phosphotyrosine interaction domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100818.1 | 733 | Silent Mutation | AAC,AAT | N,N 145 | NP_001094288.1 | |
NM_017933.4 | 733 | Silent Mutation | AAC,AAT | N,N 176 | NP_060403.3 | |
XM_011511384.2 | 733 | Silent Mutation | AAC,AAT | N,N 178 | XP_011509686.1 | |
XM_017004404.1 | 733 | Silent Mutation | AAC,AAT | N,N 233 | XP_016859893.1 | |
XM_017004405.1 | 733 | Silent Mutation | AAC,AAT | N,N 196 | XP_016859894.1 | |
XM_017004406.1 | 733 | Silent Mutation | AAC,AAT | N,N 131 | XP_016859895.1 | |
XM_017004407.1 | 733 | Silent Mutation | AAC,AAT | N,N 96 | XP_016859896.1 |