Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGAACCAATGGTTTTTTTCTTC[A/T]TGTTTCTTTTCTTTTCTTTCACTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606086 MIM: 606134 | ||||||||||||||||||||
Literature Links: |
EIF5B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF5B - eukaryotic translation initiation factor 5B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REV1 - REV1, DNA directed polymerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037872.2 | 2835 | Missense Mutation | AAG,ATG | K,M 1074 | NP_001032961.1 | |
NM_001321454.1 | 2835 | Missense Mutation | AAG,ATG | K,M 1111 | NP_001308383.1 | |
NM_001321455.1 | 2835 | Missense Mutation | AAG,ATG | K,M 1004 | NP_001308384.1 | |
NM_001321458.1 | 2835 | Missense Mutation | AAG,ATG | K,M 585 | NP_001308387.1 | |
NM_001321459.1 | 2835 | Missense Mutation | AAG,ATG | K,M 552 | NP_001308388.1 | |
NM_001321460.1 | 2835 | Missense Mutation | AAG,ATG | K,M 552 | NP_001308389.1 | |
NM_016316.3 | 2835 | Missense Mutation | AAG,ATG | K,M 1075 | NP_057400.1 | |
XM_011511341.2 | 2835 | Missense Mutation | AAG,ATG | K,M 585 | XP_011509643.1 | |
XM_017004309.1 | 2835 | Missense Mutation | AAG,ATG | K,M 1121 | XP_016859798.1 | |
XM_017004310.1 | 2835 | Missense Mutation | AAG,ATG | K,M 1111 | XP_016859799.1 | |
XM_017004311.1 | 2835 | Missense Mutation | AAG,ATG | K,M 1111 | XP_016859800.1 | |
XM_017004312.1 | 2835 | Missense Mutation | AAG,ATG | K,M 1110 | XP_016859801.1 | |
XM_017004313.1 | 2835 | Missense Mutation | AAG,ATG | K,M 1182 | XP_016859802.1 | |
XM_017004314.1 | 2835 | Missense Mutation | AAG,ATG | K,M 585 | XP_016859803.1 | |
XM_017004315.1 | 2835 | Missense Mutation | AAG,ATG | K,M 582 | XP_016859804.1 |