Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCTGAGCCACGTATGGGTGCA[C/T]GCACTGGACCTGGGGGCAGTCTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605991 | ||||||||||||||||||||
Literature Links: |
C2orf82 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2orf82 - chromosome 2 open reading frame 82 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NGEF - neuronal guanine nucleotide exchange factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114090.1 | 1109 | Missense Mutation | ATG,GTG | M,V 528 | NP_001107562.1 | |
NM_019850.2 | 1109 | Missense Mutation | ATG,GTG | M,V 620 | NP_062824.2 | |
XM_011510923.2 | 1109 | Missense Mutation | ATG,GTG | M,V 620 | XP_011509225.1 | |
XM_011510924.1 | 1109 | Missense Mutation | ATG,GTG | M,V 343 | XP_011509226.1 | |
XM_011510925.1 | 1109 | Missense Mutation | ATG,GTG | M,V 343 | XP_011509227.1 |