Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTACATGCCCTCAGACATTCTTG[C/T]TTGGAAGTAAAATTGTTTTCATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 152310 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFPI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TFPI - tissue factor pathway inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005246818.1 | 1106 | Silent Mutation | AAA,AAG | K,K 260 | XP_005246875.1 | |
XM_005246819.1 | 1106 | Silent Mutation | AAA,AAG | K,K 260 | XP_005246876.1 | |
XM_006712720.3 | 1106 | Silent Mutation | AAA,AAG | K,K 260 | XP_006712783.1 | |
XM_011511707.2 | 1106 | Silent Mutation | AAA,AAG | K,K 260 | XP_011510009.1 | |
XM_011511708.1 | 1106 | Silent Mutation | AAA,AAG | K,K 260 | XP_011510010.1 | |
XM_011511709.2 | 1106 | Silent Mutation | AAA,AAG | K,K 260 | XP_011510011.1 |