Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTATACTAGTTTCAGCAACTAAA[C/G]AGTCAGAAGTAATCCCTGTGTATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616174 | ||||||||||||||||||||
Literature Links: |
CKAP2L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CKAP2L - cytoskeleton associated protein 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304361.1 | 1917 | Missense Mutation | TCT,TGT | S,C 448 | NP_001291290.1 | |
NM_152515.4 | 1917 | Missense Mutation | TCT,TGT | S,C 613 | NP_689728.3 | |
XM_011510666.2 | 1917 | Missense Mutation | TCT,TGT | S,C 448 | XP_011508968.1 |
NT5DC4 - 5'-nucleotidase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011512262.2 | 1917 | Intron | XP_011510564.1 | |||
XM_011512265.2 | 1917 | Intron | XP_011510567.1 | |||
XM_017005474.1 | 1917 | Intron | XP_016860963.1 | |||
XM_017005475.1 | 1917 | Intron | XP_016860964.1 | |||
XM_017005476.1 | 1917 | Intron | XP_016860965.1 | |||
XM_017005477.1 | 1917 | Intron | XP_016860966.1 | |||
XM_017005478.1 | 1917 | Intron | XP_016860967.1 |