Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGACGCCCCAACTTAAGAGTCTC[A/C]TAAGTGGGGACCAGATTCTGGGAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 186854 | |||||||||||||||||||||||
Literature Links: |
GRIP2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GRIP2 - glutamate receptor interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080423.3 | 3793 | UTR 3 | NP_001073892.3 | |||
XM_011534141.2 | 3793 | UTR 3 | XP_011532443.1 | |||
XM_011534142.2 | 3793 | UTR 3 | XP_011532444.1 | |||
XM_011534143.2 | 3793 | UTR 3 | XP_011532445.1 | |||
XM_011534145.2 | 3793 | UTR 3 | XP_011532447.1 | |||
XM_011534146.2 | 3793 | UTR 3 | XP_011532448.1 |
SLC6A6 - solute carrier family 6 member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |