Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAATAACATCATATTGTAATTCCT[A/G]CTGAATACTTTTGCTCATAAGAGTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 612477 | |||||||||||||||||||||||
Literature Links: |
IQCG PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
IQCG - IQ motif containing G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134435.2 | 2173 | UTR 3 | NP_001127907.1 | |||
NM_001323027.1 | 2173 | UTR 3 | NP_001309956.1 | |||
NM_001323028.1 | 2173 | UTR 3 | NP_001309957.1 | |||
NM_001323029.1 | 2173 | UTR 3 | NP_001309958.1 | |||
NM_001323030.1 | 2173 | Intron | NP_001309959.1 | |||
NM_032263.4 | 2173 | UTR 3 | NP_115639.1 | |||
XM_017007316.1 | 2173 | Intron | XP_016862805.1 | |||
XM_017007317.1 | 2173 | Intron | XP_016862806.1 |
LRCH3 - leucine rich repeats and calponin homology domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032773.3 | 2173 | Intron | NP_116162.1 | |||
XM_005269362.2 | 2173 | Intron | XP_005269419.1 | |||
XM_005269363.2 | 2173 | Intron | XP_005269420.1 | |||
XM_005269365.2 | 2173 | Intron | XP_005269422.1 | |||
XM_005269367.2 | 2173 | Intron | XP_005269424.1 | |||
XM_006713791.3 | 2173 | Intron | XP_006713854.1 | |||
XM_006713792.3 | 2173 | Intron | XP_006713855.1 | |||
XM_006713793.2 | 2173 | Intron | XP_006713856.1 | |||
XM_011513242.2 | 2173 | Intron | XP_011511544.1 | |||
XM_011513243.2 | 2173 | Intron | XP_011511545.1 | |||
XM_017007350.1 | 2173 | Intron | XP_016862839.1 | |||
XM_017007351.1 | 2173 | Intron | XP_016862840.1 | |||
XM_017007352.1 | 2173 | Intron | XP_016862841.1 |